Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108215.1 | NP_001101685.1 | 6 | 943 | ||
XM_006244823.2 | XP_006244885.1 | 6 | 935 | |||
XM_006244824.2 | XP_006244886.1 | 6 | 1079 |