Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007198.3 | NP_009129.1 | 8 | 791 | ||
GenBank mRNA | AK021923.1 | - | 8 | 659 | ||
AK291135.1 | - | 8 | 729 | |||
AL136616.1 | - | 8 | 771 | |||
BC012334.1 | - | 8 | 781 | 3546307 | ||
CR533532.1 | - | 8 | 714 |