Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108362.2 | NP_001101832.1 | 3 | 292 | ||
XM_006251099.2 | XP_006251161.1 | 3 | 481 | |||
GenBank mRNA | BC167007.1 | - | 3 | 292 | 7310387 | |
FQ211361.1 | - | 3 | 304 | |||
FQ213588.1 | - | 3 | 325 | |||
FQ217844.1 | - | 3 | 319 | |||
FQ233251.1 | - | 3 | 296 |