Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253167.2 | XP_006253229.1 | 5 | 476 | ||
XM_341433.6 | XP_341434.3 | 5 | 473 | |||
GenBank mRNA | FQ230438.1 | - | 2 | 99 |