Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPPED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001584.2 | NP_001575.1 | 6 | 901 | ||
XM_005253110.3 | XP_005253167.1 | 7 | 1574 | |||
XM_005253111.2 | XP_005253168.1 | 7 | 1211 | |||
XM_005253112.1 | XP_005253169.1 | 7 | 1039 | |||
XM_011520346.1 | XP_011518648.1 | 8 | 1078 | |||
XM_011520347.2 | XP_011518649.1 | 7 | 1490 | |||
XM_017018231.1 | XP_016873720.1 | 8 | 1060 | |||
XM_017018233.1 | XP_016873722.1 | 8 | 1597 | |||
XM_017018234.1 | XP_016873723.1 | 8 | 1709 | |||
XM_017018235.1 | XP_016873724.1 | 8 | 1672 | |||
XM_017018236.1 | XP_016873725.1 | 6 | 1392 | |||
XM_017018237.1 | XP_016873726.1 | 6 | 972 | |||
GenBank mRNA | AK309809.1 | - | 8 | 1078 | ||
AK314833.1 | - | 9 | 1572 | |||
BC031582.1 | - | 6 | 917 | 5185528 | ||
U57911.1 | - | 6 | 901 |