Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108532.1 | NP_001102002.1 | 228 | |||
XM_006256376.2 | XP_006256438.1 | 3 | 1883 | |||
XM_006256377.2 | XP_006256439.1 | 3 | 1822 | |||
XM_006256378.2 | XP_006256440.1 | 3 | 1802 | |||
XM_006256379.2 | XP_006256441.1 | 3 | 2462 | |||
XM_006256380.2 | XP_006256442.1 | 2 | 615 |
Gene Symbol: LOC100910795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751958.3 | XP_003752006.2 | 2 | 622 | ||
XM_006256396.2 | XP_006256458.1 | 2 | 622 |