Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007204.4 | NP_009135.4 | 5, 6 | 1188 | ||
GenBank mRNA | AF106019.1 | - | 5, 6 | 841 | ||
AF155803.1 | - | 5, 6 | 836 | |||
AF171063.1 | - | 5, 6 | 1124 | |||
AK301697.1 | - | 4, 5 | 689 | |||
AK314312.1 | - | 5, 6 | 833 | |||
AL133598.1 | - | 2 | 481 | |||
BC011556.1 | - | 5, 6 | 841 | 4651697 | ||
BC031062.1 | - | 5, 6 | 850 | 5273709 | ||
BC034953.2 | - | 5, 6 | 1159 | 4824324 |