Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctex1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135030.1 | NP_001128502.1 | 5 | 565 | ||
XM_006238459.2 | XP_006238521.1 | 5 | 1321 | |||
XM_008763888.1 | XP_008762110.1 | 4 | 392 | |||
XM_008763889.1 | XP_008762111.1 | 5 | 3238 | |||
XM_008763890.1 | XP_008762112.1 | 4 | 1099 | |||
XM_008763891.1 | XP_008762113.1 | 4 | 542 |