Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007208.3 | NP_009139.1 | 10 | 1119 | ||
GenBank mRNA | AK301377.1 | - | 10 | 953 | ||
BC003375.2 | - | 10 | 1018 | 2901120 | ||
CR456720.1 | - | 10 | 924 | |||
X06323.1 | - | 10 | 1000 |