Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766787.1 | XP_008765009.1 | 7 | 921 | ||
XM_008766788.1 | XP_008765010.1 | 7 | 890 | |||
XM_008766789.1 | XP_008765011.1 | 7 | 918 |