Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007208.3 | NP_009139.1 | 5 | 714 | ||
GenBank mRNA | AK296626.1 | - | 6 | 764 | ||
AK301377.1 | - | 5 | 548 | |||
BC003375.2 | - | 5 | 613 | 2901120 | ||
BI490082.1 | - | 5 | 683 | |||
CR456720.1 | - | 5 | 519 | |||
X06323.1 | - | 5 | 595 |