Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108799.2 | NP_001102269.1 | 6 | 907 | ||
GenBank mRNA | BC168738.1 | - | 6 | 907 | 6918011 | |
FQ211594.1 | - | 6 | 703 | |||
FQ216958.1 | - | 6 | 675 | |||
FQ217523.1 | - | 6 | 675 | |||
FQ218320.1 | - | 6 | 675 | |||
FQ221159.1 | - | 6 | 675 | |||
FQ221342.1 | - | 6 | 677 | |||
FQ221482.1 | - | 6 | 675 | |||
FQ222681.1 | - | 6 | 675 | |||
FQ222690.1 | - | 6 | 676 | |||
FQ223187.1 | - | 6 | 675 | |||
FQ228255.1 | - | 6 | 676 | |||
FQ228375.1 | - | 6 | 677 | |||
FQ228595.1 | - | 6 | 676 | |||
FQ229059.1 | - | 6 | 677 | |||
FQ232283.1 | - | 6 | 679 | |||
FQ232532.1 | - | 6 | 678 |