Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007210.3 | NP_009141.2 | 6 | 1351 | ||
XM_005268607.1 | XP_005268664.1 | 6 | 1610 | |||
XM_006719214.2 | XP_006719277.1 | 5 | 1185 | |||
XM_011537819.2 | XP_011536121.1 | 7 | 1566 | |||
XM_011537822.2 | XP_011536124.1 | 6 | 1287 | |||
XM_017018744.1 | XP_016874233.1 | 6 | 1424 | |||
XM_017018745.1 | XP_016874234.1 | 7 | 1409 | |||
GenBank mRNA | AK074658.1 | - | 5 | 1249 | ||
AK296852.1 | - | 7 | 1257 | |||
BC035822.1 | - | 6 | 1354 | 5552059 | ||
BC114505.1 | - | 4 | 1030 | 40006926 |