Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047107.1 | NP_001040572.1 | 1 | 190 | ||
GenBank mRNA | DQ272745.1 | - | 1 | 190 |