Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126080.2 | NP_001119552.1 | 6 | 790 | ||
GenBank mRNA | BC091395.1 | - | 6 | 726 | 7323172 | |
FM099369.1 | - | 1 | 352 | |||
FQ229705.1 | - | 6 | 761 |