Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164746.1 | NP_001158218.1 | 3 | 671 | ||
NM_001164747.1 | NP_001158219.1 | 4 | 416 | |||
NM_001164748.1 | NP_001158220.1 | 3 | 350 | |||
NM_007211.4 | NP_009142.2 | 4 | 444 | |||
XM_006719027.2 | XP_006719090.1 | 3 | 218 | |||
XM_006719028.1 | XP_006719091.1 | 3 | 622 | |||
XM_017018746.1 | XP_016874235.1 | 3 | 258 | |||
GenBank mRNA | AB093206.1 | - | 3 | 323 | ||
AK290055.1 | - | 4 | 416 | |||
AK292333.1 | - | 3 | 665 | |||
AY665468.1 | - | 3 | 273 | |||
BC030021.1 | - | 4 | 446 | 5266519 | ||
CR456905.1 | - | 2 | 124 | |||
DB044325.1 | - | 3 | 311 | |||
U82396.1 | - | 2 | 246 |