Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ005896.1 | - | 3 | 777 | ||
AK023433.1 | - | 3 | 776 | |||
BC021213.2 | - | 3 | 763 | 4297902 |