Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000959.1 | NP_001000959.1 | 1 | 512 |
Gene Symbol: LOC100910667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766112.1 | XP_008764334.1 | 512 |
Gene Symbol: Olr1303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000796.1 | NP_001000796.1 | 1 | 512 |
Gene Symbol: Olr1304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000464.1 | NP_001000464.1 | 1 | 512 |