Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007214.4 | NP_009145.1 | 2 | 448 | ||
XM_011535399.2 | XP_011533701.1 | 2 | 197 | |||
XM_017010218.1 | XP_016865707.1 | 2 | 281 | |||
GenBank mRNA | AF100141.1 | - | 2 | 179 | ||
AJ011779.1 | - | 2 | 277 | |||
AK001465.1 | - | 2 | 311 | |||
AK074835.1 | - | 2 | 264 | |||
BC023598.2 | - | 2 | 359 | |||
BC047221.1 | - | 2 | 317 | 5277083 | ||
BC048287.1 | - | 2 | 304 | 5212007 | ||
BM764168.1 | - | 2 | 324 | |||
DB049187.1 | - | 2 | 470 | |||
KU178551.1 | - | 2 | 179 | |||
KU178552.1 | - | 2 | 179 |