Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007015.1 | NP_001007016.1 | 351 | |||
GenBank mRNA | AB005291.1 | - | 351 |