Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS5 SIRNA Assays ›
Gene Symbol:
HPS5
Gene Name:
HPS5, biogenesis of lysosomal organelles complex 2 subunit 2
Gene Aliases:
AIBP63, BLOC2S2
Chromosome Location:
Chr.11: 18278670 - 18322498 on Build GRCh38
UniGene ID:
Hs.437599
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22172
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007216.3
NP_009147.3
16, 17
2683
NM_181507.1
NP_852608.1
17, 18
2840
NM_181508.1
NP_852609.1
16, 17
2496
XM_011519862.1
XP_011518164.1
17, 18
2831
XM_011519863.1
XP_011518165.1
17, 18
2739
XM_011519864.1
XP_011518166.1
17, 18
2671
XM_011519865.1
XP_011518167.1
16, 17
3053
XM_011519866.1
XP_011518168.1
16, 17
2704
XM_011519867.1
XP_011518169.1
16, 17
2570
XM_011519868.2
XP_011518170.1
16, 17
2811
XM_017017149.1
XP_016872638.1
17, 18
2739
XM_017017150.1
XP_016872639.1
17, 18
2671
XM_017017151.1
XP_016872640.1
16, 17
2889
XM_017017152.1
XP_016872641.1
16, 17
3053
XM_017017153.1
XP_016872642.1
16, 17
2889
XM_017017154.1
XP_016872643.1
16, 17
2570
GenBank mRNA
AB023234.1
-
16, 17
2496
AF534400.1
-
16, 17
2496
AF534401.1
-
17, 18
2840
AF534402.1
-
16, 17
2683
AJ131721.1
-
5, 6
1014
AK024320.1
-
2, 3
623
AK291663.1
-
17, 18
2806
AK292436.1
-
16, 17
2649
BC033640.1
-
16, 17
2644
5502950
Back To Top
More Information
Gene Ontology Categories:
Function(s)
organelle organization
blood coagulation
pigmentation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-4h6qx:80/100.66.76.46:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline