Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008947.1 | NP_001008947.1 | 1 | 503 | ||
GenBank mRNA | AY510347.1 | - | 1 | 503 |
Gene Symbol: Vom1r64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167554.1 | NP_001161026.1 | 1 | 503 |