Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008953.1 | NP_001008953.1 | 1 | 347 | ||
GenBank mRNA | AY510352.1 | - | 1 | 347 | ||
AY510356.1 | - | 347 |
Gene Symbol: LOC100911644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758880.1 | XP_008757102.1 | 1 | 347 |
Gene Symbol: Vom1r27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008952.1 | NP_001008952.1 | 1 | 347 | ||
GenBank mRNA | AY510351.1 | - | 1 | 347 |
Gene Symbol: Vom1r29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008946.1 | NP_001008946.1 | 1 | 347 | ||
GenBank mRNA | AY510346.1 | - | 1 | 347 |