Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115030.1 | NP_001108502.1 | 2, 3 | 480 | ||
GenBank mRNA | BC091235.1 | - | 2, 3 | 458 | ||
BC158886.1 | - | 2, 3 | 480 | 8372488 |