Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007217.3 | NP_009148.2 | 3 | 447 | ||
NM_145859.1 | NP_665858.1 | 3 | 296 | |||
NM_145860.1 | NP_665859.1 | 2 | 202 | |||
XM_005247086.4 | XP_005247143.1 | 3 | 1182 | |||
XM_005247087.4 | XP_005247144.1 | 3 | 1187 | |||
XM_005247088.3 | XP_005247145.1 | 2 | 278 | |||
XM_006713485.3 | XP_006713548.1 | 3 | 342 | |||
XM_011512368.2 | XP_011510670.1 | 4 | 1255 | |||
XM_011512369.2 | XP_011510671.1 | 4 | 1260 | |||
XM_017005644.1 | XP_016861133.1 | 3 | 337 | |||
XM_017005645.1 | XP_016861134.1 | 2 | 1050 | |||
GenBank mRNA | AF022385.1 | - | 2 | 202 | ||
AK291130.1 | - | 2 | 233 | |||
BC002506.1 | - | 3 | 459 | 3050540 | ||
BC016353.1 | - | 3 | 300 | 4092244 | ||
CR457107.1 | - | 1 | 49 |