Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134337.2 | NP_001127809.1 | 4, 5 | 386 | ||
NM_001134338.2 | NP_001127810.1 | 5, 6 | 455 | |||
NM_001321749.1 | NP_001308678.1 | 4, 5 | 622 | |||
NM_007219.4 | NP_009150.1 | 4, 5 | 678 | |||
XM_011529145.2 | XP_011527447.1 | 6, 7 | 701 | |||
XM_011529146.2 | XP_011527448.1 | 5, 6 | 631 | |||
XM_011529147.2 | XP_011527449.1 | 6, 7 | 709 | |||
XM_017027620.1 | XP_016883109.1 | 5, 6 | 723 | |||
XM_017027621.1 | XP_016883110.1 | 5, 6 | 492 | |||
XM_017027622.1 | XP_016883111.1 | 5, 6 | 567 | |||
GenBank mRNA | AL079313.1 | - | 2, 3 | 94 | ||
AL096778.1 | - | 4, 5 | 476 | |||
BC000213.2 | - | 5, 6 | 409 | 2958640 | ||
BC039584.1 | - | 4, 5 | 305 | 5214313 | ||
BT007406.1 | - | 3, 4 | 224 | |||
CV811405.1 | - | 4, 5 | 364 |