Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106019.1 | NP_001099489.1 | 5 | 606 | ||
XM_006251387.2 | XP_006251449.1 | 5 | 944 | |||
XM_006251388.2 | XP_006251450.1 | 4 | 837 |
Gene Symbol: LOC685069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237209.2 | XP_006237271.1 | 1 | 532 |
Gene Symbol: RGD1560813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751821.2 | XP_003751869.1 | 1 | 560 |