Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230922.2 | XP_006230984.1 | 5 | 568 | ||
XM_006230923.2 | XP_006230985.1 | 5 | 828 | |||
XM_006230924.2 | XP_006230986.1 | 5 | 625 | |||
XM_006230925.2 | XP_006230987.1 | 5 | 592 | |||
XM_008760190.1 | XP_008758412.1 | 5 | 471 | |||
XM_574595.6 | XP_574595.3 | 5 | 1149 |