Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FM045317.1 | - | 3 | 199 | ||
FM097063.1 | - | 3 | 217 | |||
FM098643.1 | - | 3 | 389 | |||
FM110991.1 | - | 3 | 192 | |||
FQ212263.1 | - | 3 | 213 | |||
FQ220597.1 | - | 3 | 337 | |||
FQ230677.1 | - | 3 | 213 | |||
FQ230710.1 | - | 3 | 213 | |||
FQ231880.1 | - | 3 | 211 | |||
FQ234947.1 | - | 3 | 210 |