Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC500331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024337.1 | NP_001019508.1 | 4 | 720 | ||
XM_006237415.2 | XP_006237477.1 | 3 | 419 | |||
XM_008763321.1 | XP_008761543.1 | 2 | 611 | |||
XM_008763322.1 | XP_008761544.1 | 3 | 507 | |||
GenBank mRNA | BC086997.1 | - | 4 | 720 | 7131139 |
Gene Symbol: LOC100910725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237437.2 | XP_006237499.1 | 3 | 291 | ||
XM_006237438.2 | XP_006237500.1 | 206 | ||||
XM_008763340.1 | XP_008761562.1 | 611 |