Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007224.3 | NP_009155.1 | 2 | 885 | ||
GenBank mRNA | AF043469.1 | - | 1 | 38 | ||
AK094231.1 | - | 3 | 1090 | |||
AK290949.1 | - | 2 | 823 | |||
AY358614.1 | - | 2 | 876 | |||
BC036679.1 | - | 2 | 879 | 5264190 | ||
BC053581.1 | - | 2 | 809 | 5174266 |