Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192016.1 | NP_001178945.1 | 6 | 411 | ||
XM_006240179.2 | XP_006240241.1 | 6 | 724 | |||
XM_006240180.2 | XP_006240242.1 | 6 | 728 |