Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017359.2 | NP_059055.2 | 5 | 511 | ||
GenBank mRNA | AY310140.1 | - | 3 | 275 | ||
AY724493.1 | - | 5 | 690 | |||
BC085744.1 | - | 5 | 511 | 7191921 | ||
M83677.1 | - | 5 | 663 |