Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007225.2 | NP_009156.2 | 2 | 1468 | ||
GenBank mRNA | AB032985.1 | - | 1 | 1092 | ||
AL834282.1 | - | 2 | 1546 | |||
AY358526.1 | - | 2 | 1238 | |||
BC022541.1 | - | 2 | 1287 | 4794968 |