Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037504.1 | NP_001032593.1 | 2 | 96 | ||
XM_006253372.2 | XP_006253434.1 | 3 | 646 | |||
GenBank mRNA | AY621346.1 | - | 2 | 96 |