Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139060.3 | NP_620691.2 | 6 | 1093 | ||
XM_008768511.1 | XP_008766733.1 | 6 | 1098 | |||
GenBank mRNA | AB063114.1 | - | 6 | 1085 |