Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016219.4 | NP_057303.2 | 7 | 1013 | ||
NR_045720.1 | 7 | 1013 | ||||
NR_045721.1 | 8 | 1144 | ||||
XM_006716945.3 | XP_006717008.1 | 7 | 1078 | |||
XM_017014239.1 | XP_016869728.1 | 8 | 990 | |||
GenBank mRNA | AF145732.1 | - | 7 | 930 | ||
AF148509.1 | - | 7 | 990 | |||
AK074699.1 | - | 5 | 588 | |||
AK130143.1 | - | 2 | 157 | |||
AK298480.1 | - | 8 | 990 | |||
AK299043.1 | - | 7 | 860 | |||
AK309808.1 | - | 8 | 990 | |||
AK315797.1 | - | 7 | 955 | |||
AY358465.1 | - | 7 | 962 | |||
BC002953.1 | - | 7 | 946 | 3533651 | ||
BC006079.1 | - | 7 | 955 | 3959196 | ||
CR749534.1 | - | 8 | 986 |