Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191072.1 | NP_001178001.1 | 11 | 1520 | ||
XM_008762383.1 | XP_008760605.1 | 11 | 1604 | |||
XM_008762384.1 | XP_008760606.1 | 11 | 2395 | |||
XM_008762385.1 | XP_008760607.1 | 11 | 2783 |