Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007232.2 | NP_009163.2 | 3 | 2252 | ||
XM_005260266.3 | XP_005260323.1 | 4 | 1352 | |||
XM_017027623.1 | XP_016883112.1 | 4 | 1228 | |||
GenBank mRNA | AB019000.1 | - | 4 | 1352 | ||
AF140538.1 | - | 3 | 2254 | |||
AF321913.1 | - | 5 | 1112 | |||
AF346904.1 | - | 5 | 920 | |||
AK074730.1 | - | 4 | 1484 |
Set Membership: |
G-protein coupled receptor |