Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007238.4 | NP_009169.3 | 4 | 1176 | ||
NM_183397.2 | NP_899634.1 | 3 | 977 | |||
GenBank mRNA | AF072864.1 | - | 4 | 1098 | ||
AK023879.1 | - | 4 | 1094 | |||
AK297018.1 | - | 3 | 966 | |||
AL136648.1 | - | 4 | 1147 | |||
BC001147.1 | - | 4 | 1077 | 3533572 |