Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007238.4 | NP_009169.3 | 2 | 283 | ||
NM_183397.2 | NP_899634.1 | 2 | 283 | |||
GenBank mRNA | AF072864.1 | - | 2 | 205 | ||
AK023879.1 | - | 2 | 201 | |||
AK297018.1 | - | 2 | 272 | |||
AK301532.1 | - | 2 | 263 | |||
AK316589.1 | - | 2 | 263 | |||
AL136648.1 | - | 2 | 254 | |||
BC001147.1 | - | 2 | 184 | 3533572 | ||
BQ278083.1 | - | 2 | 251 | |||
CR533471.1 | - | 2 | 160 | |||
DB495125.1 | - | 2 | 283 |