Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007238.4 | NP_009169.3 | 4 | 1218 | ||
NM_183397.2 | NP_899634.1 | 3 | 1019 | |||
GenBank mRNA | AF072864.1 | - | 4 | 1140 | ||
AK023879.1 | - | 4 | 1136 | |||
AL136648.1 | - | 4 | 1189 | |||
BC001147.1 | - | 4 | 1119 | 3533572 |