Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235627.2 | XP_006235689.2 | 4 | 703 |
Gene Symbol: Eppin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GQ406385.1 | - | 3 | 406 |