Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007240.2 | NP_009171.1 | 5 | 761 | ||
XM_005244862.2 | XP_005244919.1 | 5 | 850 | |||
GenBank mRNA | AF119226.1 | - | 5 | 738 | ||
AK000259.1 | - | 3 | 175 | |||
BC006286.1 | - | 5 | 759 | 3958403 | ||
BT006633.1 | - | 5 | 727 |