Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109484.1 | NP_001102954.1 | 5 | 442 | ||
XM_008765039.1 | XP_008763261.1 | 5 | 436 | |||
XM_008765040.1 | XP_008763262.1 | 5 | 436 | |||
GenBank mRNA | BC166586.1 | - | 5 | 407 | 7934414 | |
FQ217612.1 | - | 5 | 403 | |||
FQ221005.1 | - | 5 | 413 | |||
FQ221192.1 | - | 5 | 409 | |||
FQ221499.1 | - | 5 | 411 | |||
FQ221709.1 | - | 5 | 443 | |||
FQ221718.1 | - | 5 | 414 | |||
FQ221751.1 | - | 5 | 410 | |||
FQ221963.1 | - | 5 | 411 | |||
FQ222034.1 | - | 5 | 402 | |||
FQ222160.1 | - | 5 | 412 | |||
FQ222384.1 | - | 5 | 414 | |||
FQ222504.1 | - | 5 | 391 | |||
FQ223317.1 | - | 5 | 417 | |||
FQ228533.1 | - | 5 | 414 |
Gene Symbol: Myl6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100983.1 | NP_001094453.1 | 4 | 386 | ||
GenBank mRNA | S77858.1 | - | 4 | 386 |