Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270707.1 | NP_001257636.1 | 4 | 908 | ||
NM_001270708.1 | NP_001257637.1 | 3 | 735 | |||
NM_001270709.1 | NP_001257638.1 | 3 | 713 | |||
NM_001270710.1 | NP_001257639.1 | 2 | 516 | |||
NM_007243.2 | NP_009174.1 | 5 | 1087 | |||
NR_073065.1 | 4 | 912 | ||||
NR_073066.1 | 3 | 693 | ||||
NR_073072.1 | 3 | 556 | ||||
XM_017010224.1 | XP_016865713.1 | 4 | 1089 | |||
XM_017010225.1 | XP_016865714.1 | 5 | 1003 | |||
GenBank mRNA | AF143676.1 | - | 4 | 733 | ||
AK075509.1 | - | 5 | 1089 | |||
AK298152.1 | - | 3 | 550 | |||
AK315190.1 | - | 4 | 745 | |||
AY358411.1 | - | 4 | 754 | |||
BC004509.2 | - | 2 | 359 | |||
BC021260.1 | - | 4 | 725 | |||
BC039865.1 | - | 4 | 753 | 6043663 | ||
BM917173.1 | - | 3 | 592 | |||
JQ798186.1 | - | 3 | 576 | |||
JQ798187.1 | - | 2 | 379 |