Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108528.1 | NP_001101998.1 | 1 | 92 | ||
GenBank mRNA | BC169127.1 | - | 259 | 6918056 |
Gene Symbol: RGD1561671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_341496.7 | XP_341497.4 | 1 | 51 |