Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001069267.4 | XP_001069267.2 | 3, 4 | 509 | ||
XM_006238551.2 | XP_006238613.1 | 3, 4 | 511 | |||
XM_008763922.1 | XP_008762144.1 | 3, 4 | 498 | |||
XM_008763923.1 | XP_008762145.1 | 3, 4 | 499 | |||
XM_008763924.1 | XP_008762146.1 | 3, 4 | 497 | |||
XM_008763925.1 | XP_008762147.1 | 3, 4 | 485 | |||
XM_008763926.1 | XP_008762148.1 | 3, 4 | 489 | |||
XM_008763927.1 | XP_008762149.1 | 3, 4 | 493 | |||
XM_008763929.1 | XP_008762151.1 | 3, 4 | 510 | |||
XM_008763930.1 | XP_008762152.1 | 3, 4 | 609 |