Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005892.3 | NP_005883.2 | 11 | 1236 | ||
XM_006722062.3 | XP_006722125.1 | 12 | 1451 | |||
XM_006722063.3 | XP_006722126.1 | 11 | 1431 | |||
XM_006722064.3 | XP_006722127.1 | 12 | 1421 | |||
XM_006722065.3 | XP_006722128.1 | 11 | 1401 | |||
XM_006722066.3 | XP_006722129.1 | 12 | 1447 | |||
XM_006722069.2 | XP_006722132.1 | 12 | 1450 | |||
XM_006722070.3 | XP_006722133.1 | 11 | 1427 | |||
XM_011525179.2 | XP_011523481.1 | 12 | 1451 | |||
XM_011525180.2 | XP_011523482.1 | 12 | 1143 | |||
XM_011525182.2 | XP_011523484.1 | 12 | 1448 | |||
GenBank mRNA | AF432213.1 | - | 8 | 710 | ||
AK131091.1 | - | 6 | 907 | |||
AY278319.1 | - | 11 | 1236 | |||
BC111564.1 | - | 3 | 238 | 40008718 |