Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169146.1 | NP_001162617.1 | 2 | 163 | ||
XM_006228612.2 | XP_006228674.1 | 2 | 160 | |||
GenBank mRNA | BC064030.1 | - | 137 | |||
BC084715.1 | - | 2 | 132 | |||
BC159438.1 | - | 2 | 134 | |||
FQ211832.1 | - | 2 | 144 | |||
FQ221089.1 | - | 2 | 151 | |||
FQ221637.1 | - | 2 | 145 | |||
FQ221660.1 | - | 2 | 144 | |||
FQ221692.1 | - | 2 | 142 | |||
FQ222913.1 | - | 2 | 144 | |||
FQ223100.1 | - | 2 | 145 | |||
FQ223384.1 | - | 2 | 144 | |||
FQ223396.1 | - | 2 | 145 | |||
FQ224101.1 | - | 2 | 144 | |||
FQ228433.1 | - | 2 | 145 | |||
FQ229060.1 | - | 2 | 97 | |||
X17665.1 | - | 2 | 143 |